Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentadienylthallium

Catalog No. p-7049306
Click to view available options
Quantity:
5 g
25 g

Catalog No. AA1395806

Supplier: Thermo Scientific Chemicals 01395806

Only null left
Add to Cart

CAS: 34822-90-7

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34822-90-7
MDL Number MFCD00134141
Synonym Thallium cyclopentadienide

Specifications

Melting Point (decomposition)
Physical Form Powder
Quantity 5 g
UN Number UN1707
Formula Weight 269.47
Sensitivity Air Sensitive
Chemical Name or Material Cyclopentadienylthallium
Hazard Category H300+H330-H373
Hazard Statement GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
EINECSNumber 252-229-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.