Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentadienylvanadium tetracarbonyl, 97+%

Catalog No. AA8112302
Click to view available options
Quantity:
0.5 g
2 g

Catalog No. AA8112302

Supplier: Thermo Scientific Chemicals 08112302

Only null left
Add to Cart

CAS: 12108-04-2 | C9H5O4V | 228.076 g/mol

Employed as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12108-04-2
Molecular Formula C9H5O4V
Molecular Weight (g/mol) 228.076
MDL Number MFCD00014259
InChI Key LIFGJKDEFQAOFY-UHFFFAOYSA-N
Synonym carbon monoxide; cyclopentane; vanadium, tetracarbonyl .eta.5-cyclopenta-2,4-dien-1-yl vanadium
PubChem CID 498652
IUPAC Name carbon monoxide;cyclopentane;vanadium
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[CH]1[CH][CH][CH][CH]1.[V]

Specifications

Melting Point 138°C
Boiling Point 80°C to 100°C (0.1 mmHg)(sublimation)
Quantity 0.5 g
UN Number UN3466
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 228.08
Percent Purity ≥97%
Physical Form Crystalline
Chemical Name or Material Cyclopentadienylvanadium tetracarbonyl
Hazard Category H301+H311-H315-H319-H330-H335
Hazard Statement GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
EINECSNumber 235-163-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.