Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentane, 97%, extra pure

Catalog No. p-3736067
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle

Cyclopentane, 97%, C5H10, CAS Number-287-92-3, ciclopentano, cyclopentan, cyclopentanes, cyclopentyl group, hsdb 62, hydrocarbons, cyclic c5 and c6, pentamethylene, t86pb90rnu, unii-t86pb90rnu, zyklopentan, 1L, 96% min. (GC), 05, 19, 50.0 deg.C, CHEBI:23492, White, 0.7500g/mL, 206-016-6, -7 deg.C | CAS: 287-92-3 | C5H10 | 70.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 287-92-3
Molecular Formula C5H10
Molecular Weight (g/mol) 70.15
MDL Number MFCD00001356
InChI Key RGSFGYAAUTVSQA-UHFFFAOYSA-N
Synonym pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
PubChem CID 9253
ChEBI CHEBI:23492
IUPAC Name cyclopentane
SMILES C1CCCC1

Specifications

Melting Point -94.0°C
Color White
Density 0.7500g/mL
Boiling Point 50.0°C
Flash Point −7°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4050 to 1.4070
Linear Formula C5H10
Quantity 100 mL
Residue after Evaporation 0.01 % max.
Beilstein 05, 19
Specific Gravity 0.75
Merck Index 15, 2736
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,alcohol,ether and,most hydrocarbon solvents
Viscosity 0.44 mPa.s (20°C)
Formula Weight 70.15
Percent Purity 97%
Grade Extra Pure
Physical Form Powder
Chemical Name or Material Cyclopentane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
EINECSNumber 206-016-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.