Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentane, HPLC Grade

Catalog No. p-4408383
Click to view available options
Quantity:
250 mL
1 L
4 L

For HPLC and Spectrophotometry | CAS: 287-92-3 | C5H10 | 70.135 g/mol

Cyclopentane is used in all analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 287-92-3
Molecular Formula C5H10
Molecular Weight (g/mol) 70.135
MDL Number MFCD00001356
InChI Key RGSFGYAAUTVSQA-UHFFFAOYSA-N
Synonym pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
PubChem CID 9253
ChEBI CHEBI:23492
IUPAC Name cyclopentane
SMILES C1CCCC1

Specifications

Melting Point °C to 95°C
Color Colorless
Density 0.748 g/cm3 at 20°C
Boiling Point 47°C to 49°C
Flash Point -37°C (-34°F)
Ignition Point 380°C (716°F)
Odor Petrol-like
Refractive Index 1.406
Quantity 250 mL
Beilstein 1900195
Solubility Information Miscible with ethanol,ether and acetone. Slightly miscible with water.
Vapor Pressure 398mm Hg at 20°C
Formula Weight 70.14
Grade HPLC
Physical Form Liquid
Chemical Name or Material Cyclopentane, Contains 75+% cyclopentane. Remainder consists of C5 and C6 hydrocarbons .
Show More Show Less
Hazard Category H225-H304-H335-H336
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 206-016-6
RTECSNumber GY2390000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.