Learn More
Cyclopentyl methyl ether, stab., 99.9+%
Description
Cyclopentyl methyl ether is a hydrophobic ether solvent widely used in Grignard reaction, Friedel-Crafts reaction, Claisen condensation and Beckmann reaction. It plays an important role in extraction and polymerization reactions as well as in surface coatings. It is considered as a replacement for tetrahydrofuran, methyl tert-butyl ether, dioxane and other existing ether solvents due to its excellent properties like high boiling point and relative stability under acidic and basic conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cyclopentyl methyl ether is a hydrophobic ether solvent widely used in Grignard reaction, Friedel-Crafts reaction, Claisen condensation and Beckmann reaction. It plays an important role in extraction and polymerization reactions as well as in surface coatings. It is considered as a replacement for tetrahydrofuran, methyl tert-butyl ether, dioxane and other existing ether solvents due to its excellent properties like high boiling point and relative stability under acidic and basic conditions.
Solubility
Miscible with water, alcohols, ketones and hydrocarbons.
Notes
Store in cool place. Air and light sensitive. Incompatible with oxidizing agents, acids, nitric acid, hydrogen peroxide and hydrazine.
Specifications
Specifications
| Melting Point | -140°C |
| Density | 0.86 |
| Boiling Point | 106°C |
| Flash Point | −1°C (−30°F) |
| Refractive Index | 1.421 |
| Quantity | 500 mL |
| UN Number | UN3271 |
| Solubility Information | Miscible with water,alcohols,ketones and hydrocarbons. |
| Formula Weight | 100.16 |
| Percent Purity | ≥99.9% |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H315-H319 |
| Hazard Statement | GHS H Statement H225-H302-H315-H319 Highly flammable liquid and vapor. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S. |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.