Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclophilin 40 Antibody (1B8), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB162890 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162890 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162890 Supplier Novus Biologicals Supplier No. NBP260239AF350
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cyclophilin 40 Monoclonal antibody specifically detects Cyclophilin 40 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Cyclophilin 40
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 1B8
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias 40 kDa peptidyl-prolyl cis-trans isomerase, 40 kDa peptidyl-prolyl cis-trans isomerase D, cyclophilin D, Cyclophilin-40, Cyclophilin-related protein, CYP40, CYP-40cyclophilin 40, CYPD, EC 5.2.1.8, MGC33096, peptidyl-prolyl cis-trans isomerase D, peptidylprolyl isomerase D, peptidylprolyl isomerase D (cyclophilin D), PPIase D, Rotamase D
Host Species Mouse
Immunogen Recombinant human Cyclophilin 40 (1-370aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 5481
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.