Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclophilin A/PPIA Rat anti-Human, Clone: 8H7, MilliporeSigma™

Rat Monoclonal Antibody

$182.31 - $463.74

Specifications

Antigen Cyclophilin A/PPIA
Clone 8H7
Concentration 1 mg/mL
Applications ELISA, Immunofluorescence, Immunohistochemistry, Inhibition Assays, Neutralization, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABE192425
View Documents
MilliporeSigma
MABE192425UG
25 μg
Each for $182.31
Only null left
Add to Cart
 
MABE1924100
View Documents
MilliporeSigma
MABE1924100UG
100 μg
Each for $463.74
Only null left
Add to Cart
 
Description

Description

Cyclophilin A/PPIA Monoclonal antibody specifically detects Cyclophilin A/PPIA in Human samples. It is validated for ELISA, Immunofluorescence, Immunohistochemistry, Inhibition Assay, Neutralizing, Western Blotting.
Specifications

Specifications

Cyclophilin A/PPIA
1 mg/mL
Monoclonal
Purified
RUO
Purified rat monoclonal antibody IgG2a in PBS without preservatives.
IgG2a κ
Protein G purified
8H7
ELISA, Immunofluorescence, Immunohistochemistry, Inhibition Assays, Neutralization, Western Blot
Unconjugated
Rat
Human
Ovalbumin-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of human Cyclophilin A.
Primary
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.