Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclophilin C Antibody (3C6), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N197946PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N197946PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N197946PEC5 Supplier Novus Biologicals Supplier No. NBP197946PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclophilin C Monoclonal specifically detects Cyclophilin C in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Cyclophilin C
Applications Flow Cytometry
Classification Monoclonal
Clone 3C6
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Cyclophilin C, CYPCMGC3673, EC 5.2.1.8, parvulin, peptidyl-prolyl cis-trans isomerase C, peptidylprolyl isomerase C (cyclophilin C), PPIase C, Rotamase C
Gene Symbols PPIC
Host Species Mouse
Immunogen Recombinant human PPIC (39-197aa) protein purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5480.0
Test Specificity Anti-human PPIC mAb, clone 3C6, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PPIC protein.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.