Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclophilin-E Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB057309 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB057309 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB057309 Supplier Novus Biologicals Supplier No. NBP299016H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cyclophilin-E Polyclonal specifically detects Cyclophilin-E in Human samples. It is validated for Western Blot.

Specifications

Antigen Cyclophilin-E
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias Cyclophilin E, Cyclophilin-33, CyP-33, CYP33, EC 5.2.1.8, MGC111222, MGC3736, peptidyl-prolyl cis-trans isomerase E, peptidylprolyl isomerase E (cyclophilin E), PPIase E, Rotamase E
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human Cyclophilin-E.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 10450
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.