Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclosporin A Antibody (CSZ22), Novus Biologicals™
SDP

Catalog No. NB1102569 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1102569 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1102569 Supplier Novus Biologicals Supplier No. NB1102569
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclosporin A Monoclonal specifically detects Cyclosporin A in Non-species specific samples. It is validated for ELISA.

Specifications

Antigen Cyclosporin A
Applications ELISA
Classification Monoclonal
Clone CSZ22
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Gene Alias Cyclophilin A, Cyclosporin A-binding protein, CYPAMGC12404, CYPH, EC 5.2.1.8, MGC117158, MGC23397, peptidyl-prolyl cis-trans isomerase A, peptidylprolyl isomerase A (cyclophilin A), PPIase A, Rotamase A, T cell cyclophilin
Gene Symbols PPIA
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Cyclosporin A conjugated with BSA.
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5478
Test Specificity This recognises Cyclophilin A in buffer solutions and whole blood extracts with Kd = 10^-8
Target Species Non-species specific
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.