Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYFIP1 Rabbit anti-Human, Mouse, Rat, Clone: S08-9B4, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31501925UL
This item is not returnable.
View return policy
Description
CYFIP1 Monoclonal antibody specifically detects CYFIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
CYFIP1 | |
Monoclonal | |
Unconjugated | |
cytoplasmic FMR1 interacting protein 1, cytoplasmic FMR1-interacting protein 1, FLJ45151, KIAA0068cytoplasmic FMRP interacting protein 1, P140SRA-1, selective hybridizing clone, SHYC, Specifically Rac1-associated protein 1, Sra-1, SRA1 | |
A synthetic peptide of human CYFIP1 (Uniprot#: Q7L576) | |
25 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA | |
S08-9B4 | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
Rabbit | |
Affinity-purified | |
RUO | |
23191 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction