Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYLD Antibody, Novus Biologicals™
SDP

Catalog No. NB10056461 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056461 0.1 mg
NB10056461T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056461 Supplier Novus Biologicals Supplier No. NB10056461
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

CYLD Polyclonal specifically detects CYLD in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Simple Western, Western Blot.

Specifications

Antigen CYLD
Applications Immunocytochemistry, Immunofluorescence, Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4 ug/ml, Simple Western 1:50, Immunocytochemistry/Immunofluorescence 5-10 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9NQC7
Gene Alias CDMT, CYLD1MFT, CYLDI, cylindromatosis (turban tumor syndrome), Deubiquitinating enzyme CYLD, EAC, EC 3.1.2.15, EC 3.4.19.12, FLJ20180, FLJ31664, KIAA0849FLJ78684, MFT1, probable ubiquitin carboxyl-terminal hydrolase CYLD, SBS, TEM, ubiquitin carboxyl-terminal hydrolase CYLD, ubiquitin specific peptidase like 2, ubiquitin thioesterase CYLD, Ubiquitin thiolesterase CYLD, Ubiquitin-specific-processing protease CYLD, USPL2
Gene Symbols CYLD
Host Species Rabbit
Immunogen A synthetic peptide (DKQTQKLLKVPKGS) corresponding to amino acids 32-45 of human CYLD was used as immunogen (NP_001035877).
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1540
Test Specificity Human CYLD is a 956 amino acid protein and the approximate theoretical size is 105 kD.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.