Learn More
Description
Specifications
Specifications
| Antigen | CYLN2 |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunoprecipitation 2-10 mcg/mg lysate |
| Formulation | Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide |
| Gene Accession No. | NP_003379.3 |
| Gene Alias | CAP-GLY domain containing linker protein 2, CAP-Gly domain-containing linker protein 2, CLIP, CLIP-115Williams-Beuren syndrome chromosomal region 3 protein, CYLN2WBSCR3, cytoplasmic linker 2, Cytoplasmic linker protein 115, KIAA0291Williams-Beuren syndrome chromosomal region 4 protein, WBSCR4MGC11333, Williams-Beuren syndrome chromosome region 3, Williams-Beuren syndrome chromosome region 4, WSCR3, WSCR4Cytoplasmic linker protein 2 |
| Gene Symbols | CLIP2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
