Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYLN2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB290872 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290872 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290872 Supplier Novus Biologicals Supplier No. NBP178744PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYLN2 Polyclonal antibody specifically detects CYLN2 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen CYLN2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias CAP-GLY domain containing linker protein 2, CAP-Gly domain-containing linker protein 2, CLIP, CLIP-115Williams-Beuren syndrome chromosomal region 3 protein, CYLN2WBSCR3, cytoplasmic linker 2, Cytoplasmic linker protein 115, KIAA0291Williams-Beuren syndrome chromosomal region 4 protein, WBSCR4MGC11333, Williams-Beuren syndrome chromosome region 3, Williams-Beuren syndrome chromosome region 4, WSCR3, WSCR4Cytoplasmic linker protein 2
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 996 and 1046 of human Cytoplasmic Linker Protein 115 using the numbering given in entry NP_003379.3 (GeneID 7461).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7461
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.