Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CYM 50260
GREENER_CHOICE

Catalog No. 467710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-771-0 10 mg
46-775-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-771-0 Supplier Tocris Bioscience™ Supplier No. 4677/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective S1P4 agonist

Potent sphingosine-1-phosphate receptor 4 (S1P4) agonist (EC50 = 45 nM). Displays no activity at other S1P receptor subtypes (S1P1-3 and S1P5) at concentrations up to 25 μM.

Chemical Identifiers

CAS 1355026-60-6
Molecular Formula C14H11Cl3FNO2

Specifications

Quantity 10 mg
Formula Weight 350.6
Purity >98%
Molecular Weight (g/mol) 350.595
Chemical Name or Material CYM 50260
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.