Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CYM 9484
GREENER_CHOICE

Catalog No. 460610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-061-0 10 mg
46-065-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-061-0 Supplier Tocris Bioscience™ Supplier No. 4606/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent NPY Y2 receptor antagonist

Potent neuropeptide Y (NPY) Y2 receptor antagonist (IC50 = 19 nM).

Chemical Identifiers

CAS 1383478-94-1
Molecular Formula C27H31N3O3S2

Specifications

Quantity 10 mg
Formula Weight 509.63
Purity >98%
Molecular Weight (g/mol) 509.683
Chemical Name or Material CYM 9484
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.