Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP1B1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB048942 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB048942 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB048942 Supplier Novus Biologicals Supplier No. NBP297885AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP1B1 Polyclonal specifically detects CYP1B1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CYP1B1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias aryl hydrocarbon hydroxylase, CP1B, CYPIB1, cytochrome P450 1B1, cytochrome P450, family 1, subfamily B, polypeptide 1, cytochrome P450, subfamily I (dioxin-inducible), polypeptide 1 (glaucoma 3, EC 1.14.14.1, flavoprotein-linked monooxygenase, GLC3A, microsomal monooxygenase, P4501B1, primary infantile), xenobiotic monooxygenase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CYP1B1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1545
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.