Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP1B1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB007694 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB007694 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB007694 Supplier Novus Biologicals Supplier No. NBP250551JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP1B1 Polyclonal specifically detects CYP1B1 in Human, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CYP1B1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias aryl hydrocarbon hydroxylase, CP1B, CYPIB1, cytochrome P450 1B1, cytochrome P450, family 1, subfamily B, polypeptide 1, cytochrome P450, subfamily I (dioxin-inducible), polypeptide 1 (glaucoma 3, EC 1.14.14.1, flavoprotein-linked monooxygenase, GLC3A, microsomal monooxygenase, P4501B1, primary infantile), xenobiotic monooxygenase
Gene Symbols CYP1B1
Host Species Rabbit
Immunogen This CYP1B1 Antibody was developed against a portion of amino acids 200-250 of human CYP1B1 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Target Species Human, Canine, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.