Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP1B1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224722SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224722SS 0.025 mg
NBP224722 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224722SS Supplier Novus Biologicals Supplier No. NBP224722SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

CYP1B1 Polyclonal specifically detects CYP1B1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, In vitro assay.

Specifications

Antigen CYP1B1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), In vitro Assay
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.5 to 2 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 5 to 10 μg/mL, In vitro assay
Formulation PBS with 0.05% Sodium Azide
Gene Alias aryl hydrocarbon hydroxylase, CP1B, CYPIB1, cytochrome P450 1B1, cytochrome P450, family 1, subfamily B, polypeptide 1, cytochrome P450, subfamily I (dioxin-inducible), polypeptide 1 (glaucoma 3, EC 1.14.14.1, flavoprotein-linked monooxygenase, GLC3A, microsomal monooxygenase, P4501B1, primary infantile), xenobiotic monooxygenase
Gene Symbols CYP1B1
Host Species Rabbit
Immunogen A portion of amino acids 400-450 of human CYP1B1 was used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1545
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 93% in bovine and pig, and 87% in mouse, and dog.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.