Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP1B1 Antibody (V96), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309790 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309790 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309790 Supplier Novus Biologicals Supplier No. NBP250191MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP1B1 Monoclonal antibody specifically detects CYP1B1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen CYP1B1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V96
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias aryl hydrocarbon hydroxylase, CP1B, CYPIB1, cytochrome P450 1B1, cytochrome P450, family 1, subfamily B, polypeptide 1, cytochrome P450, subfamily I (dioxin-inducible), polypeptide 1 (glaucoma 3, EC 1.14.14.1, flavoprotein-linked monooxygenase, GLC3A, microsomal monooxygenase, P4501B1, primary infantile), xenobiotic monooxygenase
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide - GSAFADRPAF.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1545
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.