Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP20A1 Antibody (Z27-P3H7*E3), FITC, Novus Biologicals™
SDP

Catalog No. NBP250192F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250192F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250192F Supplier Novus Biologicals Supplier No. NBP250192F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP20A1 Monoclonal specifically detects CYP20A1 in Human samples. It is validated for ELISA, Immunohistochemistry.

Specifications

Antigen CYP20A1
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone Z27-P3H7*E3
Conjugate FITC
Dilution ELISA, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CYP-M, cytochrome P450 20A1, cytochrome P450 monooxygenase, cytochrome P450, family 20, subfamily A, polypeptide 1, EC 1.14, MGC22229
Gene Symbols CYP20A1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide AWITVSKRY
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 57404
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.