Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP21A2 Antibody, Novus Biologicals™
SDP

Catalog No. NB404983 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB404983 25ul
NBP213893 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB404983 Supplier Novus Biologicals Supplier No. NBP21389325UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP21A2 Polyclonal specifically detects CYP21A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CYP21A2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500-1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias 21-OHase, CA21HCYP21B, CAH1, CPS1, CYP21EC 1.14.99.10, Cytochrome P450 21, Cytochrome P450 XXI, cytochrome P450, family 21, subfamily A, polypeptide 2, cytochrome P450, subfamily XXIA (steroid 21-hydroxylase, congenital adrenalhyperplasia), polypeptide 2, Cytochrome P-450c21, Cytochrome P450-C21, Cytochrome P450-C21B, EC 1.14.99, MGC150536, MGC150537, P450c21B, steroid 21-hydroxylase, steroid 21-monooxygenase
Gene Symbols CYP21A2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: GLTQKFGPIYRLHLGLQDVVVLNSKRTIEEAMVKKWADFAGRPEPLTYKLVSRNYPDLSLGDYSLLW
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1589
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.