Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP24A1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP152111 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152111 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152111 Supplier Novus Biologicals Supplier No. NBP152111
Add to Cart
Add to Cart

Goat Polyclonal Antibody

CYP24A1 Polyclonal specifically detects CYP24A1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen CYP24A1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_000773.2
Gene Alias 25-dihydroxyvitamin D(3) 24-hydroxylase, mitochondrial, CP241, CYP241,25-alphadihydroxyvitamin D3 24-hydroxylase, Cytochrome P450 24A1, cytochrome P450, family 24, subfamily A, polypeptide 1,24-OHase, cytochrome P450, subfamily XXIV (vitamin D 24-hydroxylase), Cytochrome P450-CC24, EC 1.14.13.n4, exo-mitochondrial protein, MGC126273, MGC126274, P450-CC24, vitamin D 24-hydroxylase, Vitamin D(3) 24-hydroxylase
Gene Symbols CYP24A1
Host Species Goat
Immunogen Peptide with sequence C-RKYDIQATDN corresponding to internal region according to NP_000773.2, NP_001122387.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1591
Test Specificity This antibody is expected to recognize both reported isoforms (NP_000773.2; NP_001122387.1).
Target Species Human, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.