Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP26A1 Rabbit anti-Human, Mouse, Rat, Clone: 1E1D2, Novus Biologicals™
SDP

Catalog No. NB168345 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168345 100 μg
NB168346 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168345 Supplier Novus Biologicals Supplier No. NBP316627100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CYP26A1 Monoclonal antibody specifically detects CYP26A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CYP26A1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1E1D2
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CP26, CYP26Retinoic acid 4-hydroxylase, cytochrome P450 26A1, Cytochrome P450 retinoic acid-inactivating 1, cytochrome P450, family 26, subfamily A, polypeptide 1, cytochrome P450, subfamily XXVIA, polypeptide 1, Cytochrome P450RAI, EC 1.14, hP450RAI, P450, retinoic acid-inactivating, 1, P450RAI, P450RAI1Retinoic acid-metabolizing cytochrome
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 201-450 of human CYP26A1 (O43174). AGDGDSEQQLVEAFEEMTRNLFSLPIDVPFSGLYRGMKARNLIHARIEQNIRAKICGLRASEAGQGCKDALQLLIEHSWERGERLDMQALKQSSTELLFGGHETTASAATSLITYLGLYPHVLQKVREELKSKGLLCKSNQDNKLDMEILEQLKYIGCVIKETLRLNPPVPGGFRVALKTFELNGYQIPKGWNVIYSICDTHDVAEIFTNKEEFNPDRFMLPHPEDASRFSFIPFGGGLRSCVGKEFAKI
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1592
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.