Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP26C1 Antibody (T6P1C7*E7), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP250196AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250196AH 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250196AH Supplier Novus Biologicals Supplier No. NBP250196AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP26C1 Monoclonal specifically detects CYP26C1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen CYP26C1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone T6P1C7*E7
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cytochrome P450 26C1, cytochrome P450, family 26, subfamily C, polypeptide 1, EC 1.14, FLJ45301
Gene Symbols CYP26C1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide RWELATPAFP (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340665
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.