Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP26C1 Antibody (T6P1C7*E7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB395512 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395512 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395512 Supplier Novus Biologicals Supplier No. NBP2501960.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP26C1 Monoclonal antibody specifically detects CYP26C1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen CYP26C1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone T6P1C7*E7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Formulation PBS
Gene Alias cytochrome P450 26C1, cytochrome P450, family 26, subfamily C, polypeptide 1, EC 1.14, FLJ45301
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide RWELATPAFP (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 340665
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.