Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP27A1 Antibody (V29-P4B8*B2), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP276660G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276660G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP276660G Supplier Novus Biologicals Supplier No. NBP276660G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP27A1 Monoclonal specifically detects CYP27A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CYP27A1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone V29-P4B8*B2
Conjugate DyLight 488
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 12-alpha-triol 27-hydroxylase, 7-alpha, cholestanetriol 26-monooxygenase, CP27,5-beta-cholestane-3-alpha, 7-alpha, 12-alpha-triol 27-hydroxylase, CTX5-beta-cholestane-3-alpha, CYP275-beta-cholestane-3-alpha, 7-alpha, 12-alpha-triol 26-hydroxylase, Cytochrome P450 27, cytochrome P450, family 27, subfamily A, polypeptide 1, cytochrome P450, subfamily XXVIIA (steroid 27-hydroxylase, cerebrotendinousxanthomatosis), polypeptide 1, Cytochrome P-450C27/25, EC 1.14.13.15, sterol 26-hydroxylase, mitochondrial, Sterol 27-hydroxylase, Vitamin D(3) 25-hydroxylase
Gene Symbols CYP27A1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide KVVLAPETGEL (C-terminal sequence)
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1593
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.