Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP2D6 Mouse anti-Human, Clone: 8B11D12, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | CYP2D6 |
|---|---|
| Clone | 8B11D12 |
| Concentration | 1.85 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6392
![]()
|
Proteintech Group Inc
602971IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is known to metabolize as many as 20% of commonly prescribed drugs. Its substrates include debrisoquine, an adrenergic-blocking drug; sparteine and propafenone, both anti-arrythmic drugs; and amitryptiline, an anti-depressant. The gene is highly polymorphic in the population; certain alleles result in the poor metabolizer phenotype, characterized by a decreased ability to metabolize the enzyme′s substrates. The gene is located near two cytochrome P450 pseudogenes on chromosome 22q13.1. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| CYP2D6 | |
| 1.85 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CPD6, CYP2D, CYP2D6, CYP2DL1, Cytochrome P450 2D6, P450 DB1, P450C2D, P450DB1 | |
| CYP2D6 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 8B11D12 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P10635 | |
| 1565 | |
| CYP2D6 Fusion Protein Ag12605 | |
| Primary | |
| -20°C | |
| CYP2D6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title