Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP2R1 Antibody (M26-P6H1), Biotin, Novus Biologicals™
SDP

Catalog No. NBP250204B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250204B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250204B Supplier Novus Biologicals Supplier No. NBP250204B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP2R1 Monoclonal specifically detects CYP2R1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen CYP2R1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone M26-P6H1
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Cytochrome P450 2R1, cytochrome P450, family 2, R1, cytochrome P450, family 2, subfamily R, polypeptide 1, EC 1.14.13.15, MGC4663, vitamin D 25-hydroxylase
Gene Symbols CYP2R1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide QPYLICAERR (C-terminal sequence).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 120227
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.