Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP2S1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CYP2S1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CYP2S1 Polyclonal specifically detects CYP2S1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
CYP2S1 | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
29785 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GTVAMLEGTFDGHGVFFSNGERWRQLRKFTMLALRDLGMGKREGEELIQAEARCLVETFQGTEGRPFDPSLLLAQATSNVVCSLL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
CYPIIS1, cytochrome P450 2S1, cytochrome P450 family member predicted from ESTs, cytochrome P450, family 2, subfamily S, polypeptide 1, cytochrome P450, subfamily IIS, polypeptide 1, cytochrome P540, subfamily IIS, polypeptide 1, EC 1.14.14.1 | |
CYP2S1 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title