Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP2W1 Mouse anti-Human, Biotin, Clone: V32-P5E5, Novus Biologicals™
SDP

Catalog No. NBP250205B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250205B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250205B Supplier Novus Biologicals Supplier No. NBP250205B

Mouse Monoclonal Antibody

CYP2W1 Monoclonal specifically detects CYP2W1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen CYP2W1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V32-P5E5
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CYPIIW1, cytochrome P450 2W1, cytochrome P450, family 2, subfamily W, polypeptide 1, EC 1.14.14.-, EC 1.14.14.1, EC 1.14.14.5, FLJ20359, MGC34287
Gene Symbols CYP2W1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide - RYRLLPPPGV (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54905
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.