Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP2W1 Mouse anti-Human, Janelia Fluor 549, Clone: V32-P5E5, Novus Biologicals™
SDP

Catalog No. NBP50205549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP50205549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP50205549 Supplier Novus Biologicals Supplier No. NBP250205JF549

Mouse Monoclonal Antibody

CYP2W1 Monoclonal specifically detects CYP2W1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen CYP2W1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V32-P5E5
Conjugate Janelia Fluor 549
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CYPIIW1, cytochrome P450 2W1, cytochrome P450, family 2, subfamily W, polypeptide 1, EC 1.14.14.-, EC 1.14.14.1, EC 1.14.14.5, FLJ20359, MGC34287
Gene Symbols CYP2W1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide - RYRLLPPPGV (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 54905
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.