Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP39A1 Antibody (M30P6D6), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP250206FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250206FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250206FR Supplier Novus Biologicals Supplier No. NBP250206FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP39A1 Monoclonal specifically detects CYP39A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen CYP39A1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone M30P6D6
Conjugate DyLight 680
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cytochrome P450 39A1, cytochrome P450, family 39, subfamily A, polypeptide 1, cytochrome P450, subfamily XXXIX (oxysterol 7 alpha-hydroxylase), polypeptide 1,24-hydroxycholesterol 7-alpha-hydroxylase, EC 1.14.13.99, hCYP39A1, oxysterol 7alpha-hydroxylase, Oxysterol 7-alpha-hydroxylase
Gene Symbols CYP39A1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide CRIEYKQRI (C-terminal sequence).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51302
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.