Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP3A43 Antibody, Novus Biologicals™
SDP

Catalog No. NBP216064 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216064 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216064 Supplier Novus Biologicals Supplier No. NBP216064
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP3A43 Polyclonal specifically detects CYP3A43 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen CYP3A43
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9HB55
Gene Alias cytochrome P450 3A43, cytochrome P450 polypeptide 43, cytochrome P450, family 3, subfamily A, polypeptide 43, cytochrome P450, subfamily IIIA, polypeptide 43, EC 1.14.14.1, MGC119315, MGC119316
Gene Symbols CYP3A43
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CYP3A43. The exact sequence is proprietary.
Molecular Weight of Antigen 45 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 64816
Test Specificity Human. Expected to cross react based on sequence identity: Monkey (97%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.