Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP3A43 Antibody, Novus Biologicals™
SDP

Catalog No. NBP169413 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP169413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP169413 Supplier Novus Biologicals Supplier No. NBP169413
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP3A43 Polyclonal specifically detects CYP3A43 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CYP3A43
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9HB55
Gene Alias cytochrome P450 3A43, cytochrome P450 polypeptide 43, cytochrome P450, family 3, subfamily A, polypeptide 43, cytochrome P450, subfamily IIIA, polypeptide 43, EC 1.14.14.1, MGC119315, MGC119316
Gene Symbols CYP3A43
Host Species Rabbit
Immunogen Synthetic peptides corresponding to CYP3A43(cytochrome P450, family 3, subfamily A, polypeptide 43) The peptide sequence was selected from the C terminal of CYP3A43. Peptide sequence IYALHHDPKYWTEPEKFCPESRFSKKNKDSIDLYRYIPFGAGPRNCIGMR.
Molecular Weight of Antigen 58 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 64816
Test Specificity Canine: 86%; Goat: 85%; Mouse: 85%; Guinea pig: 80%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Sheep
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.