Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP3A5 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501728970 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
Catalog No. Quantity
50-172-8970 150 μL
1 options

Catalog No. 50-172-8970

Supplier: Proteintech Group Inc 137371AP150UL

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP3A5 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by glucocorticoids and some pharmacological agents. The enzyme metabolizes drugs such as nifedipine and cyclosporine as well as the steroid hormones testosterone, progesterone and androstenedione.

Specifications

Antigen CYP3A5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CYP3A5
Gene Accession No. P20815
Gene Alias CP35, CYP3A5, CYPIIIA5, Cytochrome P450 3A5, Cytochrome P450 HLp2, Cytochrome P450 PCN3, P450PCN3, PCN3
Gene Symbols CYP3A5
Host Species Rabbit
Immunogen CYP3A5 Fusion Protein Ag4674
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1577
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.