Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP3C1 Antibody (V12P6F5*E3), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP250210UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250210UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250210UV Supplier Novus Biologicals Supplier No. NBP250210UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP3C1 Monoclonal specifically detects CYP3C1 in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen CYP3C1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V12P6F5*E3
Conjugate DyLight 350
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CYP3A, cyp3c1, Cytochrome P450, family 3, subfamily c, polypeptide 1, wu:fa96d12, wu:fc26b04, zgc:56626
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide KLIPRSHKEK (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Target Species Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.