Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP46A1 Antibody (N8-P6E4*H8), FITC, Novus Biologicals™
SDP

Catalog No. NBP250194F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250194F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250194F Supplier Novus Biologicals Supplier No. NBP250194F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP46A1 Monoclonal specifically detects CYP46A1 in Human samples. It is validated for ELISA, Immunohistochemistry.

Specifications

Antigen CYP46A1
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone N8-P6E4*H8
Conjugate FITC
Dilution ELISA, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CH24H, cholesterol 24-hydroxylase, CYP46CP46, Cytochrome P450 46A1, cytochrome P450, family 46, subfamily A, polypeptide 1, cytochrome P450, subfamily 46 (cholesterol 24-hydroxylase), EC 1.14.13.98
Gene Symbols CYP46A1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide- PVLCTLRPR (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10858
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.