Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP4B1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP132691 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP132691 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP132691 Supplier Novus Biologicals Supplier No. NBP132691
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP4B1 Polyclonal specifically detects CYP4B1 in Human samples. It is validated for Western Blot.

Specifications

Antigen CYP4B1
Applications Western Blot
Classification Polyclonal
Concentration 0.69mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias CYPIVB1, cytochrome P450 4B1, cytochrome P450, family 4, subfamily B, polypeptide 1, cytochrome P450, subfamily IVB, polypeptide 1, Cytochrome P450-HP, EC 1.14.14.1, microsomal monooxygenase, P-450HP
Gene Symbols CYP4B1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CYP4B1. The exact sequence is proprietary.
Molecular Weight of Antigen 59 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1580
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.