Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP4F11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501732654 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2654 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2654 Supplier Proteintech Group Inc Supplier No. 200121AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody is specific to CYP4F11. It has no cross reaction to other CYP4F members.

The cytochrome P450 proteins (CYPs) are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. P450 enzymes are classified into subfamilies based on their sequence similarities. CYP4F11 is an isoform of the cytochrome P450 4F protein and is expressed mainly in human liver, followed by kidney, heart, and skeletal muscle. CYP4F11 contains 524 amino acid residues that share 80.0, 82.3 and 79.2% identity to CYP4F2, CYP4F3, and CYP4F8 amino acid sequences, respectively. CYP4F11 has catalytic properties towards endogenous eicosanoids as well as some clinically relevant drugs, and is able to metabolize large molecules such as erythromycin.

Specifications

Antigen CYP4F11
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CYP4F11
Gene Accession No. Q9HBI6
Gene Alias CYP4F11, CYPIVF11, Cytochrome P450 4F11
Gene Symbols CYP4F11
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57834
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.