Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP4F12 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen CYP4F12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP162396
SDP
View Documents
Novus Biologicals
NBP162396
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

CYP4F12 Polyclonal specifically detects CYP4F12 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CYP4F12
Polyclonal
Rabbit
Lipid and Metabolism
CYPIVF12, cytochrome P450 4F12, cytochrome P450, family 4, subfamily F, polypeptide 12, cytochrome P450, subfamily IVF, polypeptide 12, EC 1.14.14.1, F22329_1
CYP4F12
IgG
60 kDa
Western Blot
Unconjugated
RUO
Q9HCS2
66002
Synthetic peptides corresponding to CYP4F12(cytochrome P450, family 4, subfamily F, polypeptide 12) The peptide sequence was selected from the C terminal of CYP4F12. Peptide sequence TVWPDPEVYDPFRFDPENSKGRSPLAFIPFSAGPRNCIGQAFAMAEMKVV.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.