Learn More
Invitrogen™ CYP4F12 Polyclonal Antibody

Description
CYP4F12 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
Specifications
| Antigen | CYP4F12 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CYP4F12 |
| Gene Accession No. | Q9HCS2 |
| Gene Alias | CYP4F12; CYPIVF12; Cytochrome P450 4F12; cytochrome P450 family 4 subfamily F member 12; cytochrome P450, family 4, subfamily F, polypeptide 12; cytochrome P450, subfamily IVF, polypeptide 12; F22329_1; UNQ568/PRO1129 |
| Gene Symbols | CYP4F12 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.