Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP4V2 Antibody (M29-P3B10), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB309752 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309752 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309752 Supplier Novus Biologicals Supplier No. NBP250214MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP4V2 Monoclonal antibody specifically detects CYP4V2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen CYP4V2
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone M29-P3B10
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias BCD, CYP4AH1, cytochrome P450 4V2, cytochrome P450, family 4, subfamily V, polypeptide 2, EC 1.14, EC 1.14.14.1, FLJ18432, MGC43534
Host Species Mouse
Immunogen Peptide sequence - KREELGLEGQ (amino acids 495 - 504)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 285440
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.