Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP4Z1 Antibody (N7-P2G5*D8), Biotin, Novus Biologicals™
SDP

Catalog No. NBP250215B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250215B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250215B Supplier Novus Biologicals Supplier No. NBP250215B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP4Z1 Monoclonal specifically detects CYP4Z1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen CYP4Z1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone N7-P2G5*D8
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CYP4A20, CYPIVZ1, cytochrome P450 4Z1, cytochrome P450, family 4, subfamily Z, polypeptide 1, EC 1.14.14.1
Gene Symbols CYP4Z1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide KLAPDHSRPP (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cell Cycle and Replication, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 199974
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.