Learn More
Description
Specifications
Specifications
| Antigen | CYP51A1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | NP_000777 |
| Gene Alias | CP51, CYP51Cytochrome P450LI, CYPL1, CYPLI, Cytochrome P450 51A1, cytochrome P450, family 51, subfamily A, polypeptide 1, Cytochrome P450-14DM, EC 1.14.13.70, lanosterol 14-alpha demethylase, lanosterol 14-alpha-demethylase, LDMCytochrome P45014DM, P450-14DM, P450L1,51 (lanosterol 14-alpha-demethylase), Sterol 14-alpha demethylase |
| Gene Symbols | CYP51A1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
