Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP7B1 Antibody (M17-P3F2), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NBP50217549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP50217549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP50217549 Supplier Novus Biologicals Supplier No. NBP250217JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP7B1 Monoclonal specifically detects CYP7B1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CYP7B1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone M17-P3F2
Conjugate Janelia Fluor 549
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CBAS3, CP7B, Cytochrome P450 7B1, cytochrome P450, family 7, subfamily B, polypeptide 1, cytochrome P450, subfamily VIIB (oxysterol 7 alpha-hydroxylase), polypeptide 1,25-hydroxycholesterol 7-alpha-hydroxylase, EC 1.14.13.100, oxysterol 7alpha-hydroxylase, Oxysterol 7-alpha-hydroxylase, spastic paraplegia 5A (autosomal recessive), SPG5A
Gene Symbols CYP7B1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide IQYPDSDVL (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9420
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.