Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyr61/CCN1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB10356A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10356A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10356A750 Supplier Novus Biologicals Supplier No. NB100356AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cyr61/CCN1 Polyclonal specifically detects Cyr61/CCN1 in Human, Mouse, Rabbit samples. It is validated for Block/Neutralize, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Cyr61/CCN1
Applications Blocking Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CCN family member 1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1
Gene Symbols CCN1
Host Species Rabbit
Immunogen A synthetic peptide made to the human CYR61 protein sequence (between residues 150-250). [UniProtNo. O00622]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 3491.0
Target Species Human, Mouse, Rabbit
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.