Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyr61/CCN1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB100356 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100356 0.1 mL
NB100356SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100356 Supplier Novus Biologicals Supplier No. NB100356
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 13 publications

Cyr61/CCN1 Polyclonal specifically detects Cyr61/CCN1 in Human, Mouse, Rabbit samples. It is validated for Block/Neutralize, Functional Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot.

Specifications

Antigen Cyr61/CCN1
Applications Blocking Assay, Functional Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:200, Simple Western 1:400, Immunohistochemistry reported in scientific literature, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 18202125), Immunohistochemistry-Paraffin 1:100, Block/Neutralize reported in scientific literature (PMID 22282654)
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. O00622
Gene Alias CCN family member 1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1
Gene Symbols CCN1
Host Species Rabbit
Immunogen A synthetic peptide made to the human CYR61 protein sequence (between residues 150-250). [UniProt# O00622]
Molecular Weight of Antigen 37 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3491
Test Specificity Human. The immunogen sequence has 88% homology to rat and mouse.
Target Species Human, Rabbit, Mouse, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.