Learn More
Description
Specifications
Specifications
| Antigen | Cyr61/CCN1 |
| Applications | Blocking Assay, Functional Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:200, Simple Western 1:400, Immunohistochemistry reported in scientific literature, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 18202125), Immunohistochemistry-Paraffin 1:100, Block/Neutralize reported in scientific literature (PMID 22282654) |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | O00622 |
| Gene Alias | CCN family member 1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1 |
| Gene Symbols | CCN1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
