Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYRPTIC/CFC1 Antibody (MM0205-8D12), FITC, Novus Biologicals™
SDP

Catalog No. NB344280 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344280 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344280 Supplier Novus Biologicals Supplier No. NBP212209F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYRPTIC/CFC1 Monoclonal antibody specifically detects CYRPTIC/CFC1 in Human samples. It is validated for Western Blot

Specifications

Antigen CYRPTIC/CFC1
Applications Western Blot
Classification Monoclonal
Clone MM0205-8D12
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias CFC1B, cripto, FRL-1, cryptic family 1, Cryptic family protein 1, cryptic protein, CRYPTICFLJ77897, HTX2MGC133213
Host Species Mouse
Immunogen Human recombinant CYRPTIC/CFC1
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55997
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.